missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB6 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76519AF532
This item is not returnable.
View return policy
Description
ZBTB6 Polyclonal antibody specifically detects ZBTB6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ZBTB6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 532 | |
| zinc finger and BTB domain containing 6, zinc finger and BTB domain-containing protein 6, Zinc finger protein 482ZIDZinc finger protein with interaction domain, ZNF482 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the center of human ZBTB6. The immunogen is located within amino acids 180 - 230 of ZBTB6. Amino Acid Squence: ESLTSERKEMKSPELS | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 10773 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction