missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZBTB6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567439
This item is not returnable.
View return policy
Description
Immunogen sequence: EGSYGTVSEIQ NLEEGYSLRH QCPRCPRGFL HVENYLRHLK MHKLFLCLQC Highest antigen sequence identity to the following orthologs - mouse 88%, rat 86%.
The ZBTB family of proteins is comprised of diverse zinc finger proteins that also contain a BTB (BR-C, ttk and bab) domain. ZBTB6, also known as ZNF482, is highly expressed in brain. While little is known about ZBTB6, other ZBTB proteins, such as ZBTB4 bind methylated DNA and repress transcription. Another ZBTB family member, ZBTB5, also acts as a transcriptional repressor, and can interact with co-repressor histone deacetylase complexes such as BCoR, NCoR, and SMRT via its POZ domain, suggesting that ZBTB6 may also act as a transcriptional repressor.
Specifications
| ZBTB6 | |
| Polyclonal | |
| Unconjugated | |
| ZBTB6 | |
| A830092L04Rik; ZBTB6; Zfp482; ZID; zinc finger and BTB domain containing 6; zinc finger and BTB domain-containing protein 6; Zinc finger protein 482; zinc finger protein with interaction domain; ZNF482 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15916 | |
| ZBTB6 | |
| Recombinant Human ZBTB6. Recombinant protein control fragment (Product #RP-108100). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction