missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB7A/Pokemon Antibody (SR2167), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | ZBTB7A/Pokemon |
|---|---|
| Clone | SR2167 |
| Dilution | Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649774
|
Novus Biologicals
NBP3-22161-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622335
|
Novus Biologicals
NBP3-22161-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZBTB7A/Pokemon Monoclonal antibody specifically detects ZBTB7A/Pokemon in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| ZBTB7A/Pokemon | |
| Western Blot 1:1000-1:5000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp547O146, Factor binding IST protein 1, Factor that binds to inducer of short transcripts protein 1, FBI-1POK erythroid myeloid ontogenic factor, FBI1TTF-I-interacting peptide 21, HIV-1 inducer of short transcripts binding protein, Leukemia/lymphoma-related factor, LRFHIV-1 1st-binding protein 1, lymphoma related factor, MGC99631, pokemon, TIP21, ZBTB7zinc finger and BTB domain containing 7, zinc finger and BTB domain containing 7A, zinc finger and BTB domain containing 7A, HIV-1 inducer of short transcriptsbinding protein, zinc finger and BTB domain-containing protein 7A, Zinc finger protein 857A, ZNF857APOZ and Krueppel erythroid myeloid ontogenic factor | |
| A synthesized peptide derived from human ZBTB7A/Pokemon (Uniprot #: O95365) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2167 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 51341 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title