missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB8A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 168.00
Specifications
| Antigen | ZBTB8A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
ZBTB8A Polyclonal specifically detects ZBTB8A in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZBTB8A | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CAI20024 | |
| 653121 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ZBTB8. The immunogen is located within the last 50 amino acids of ZBTB8. Amino Acid Squence: VDDSEEEEEKEIKPNIR | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| BOZ-F1, BOZF1BTB/POZ and zinc-finger domains factor on chromosome 1, BTB/POZ and zinc-finger domain-containing factor, FLJ90065, MGC17919, ZBTB8, zinc finger and BTB domain containing 8, zinc finger and BTB domain containing 8A, zinc finger and BTB domain-containing protein 8A, ZNF916A | |
| ZBTB8A | |
| IgG | |
| Affinity Purified | |
| At least two isoforms of ZBTB8A are known to exist; this antibody will recognize both isoforms. This antibody is predicted to not cross-react with other ZBTB protein family members. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title