missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H13 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2276B
This item is not returnable.
View return policy
Description
ZC3H13 Polyclonal antibody specifically detects ZC3H13 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ZC3H13 | |
| Polyclonal | |
| Western Blot | |
| FLJ35669, KIAA0853DKFZp434D1812, zinc finger CCCH domain-containing protein 13, zinc finger CCCH-type containing 13 | |
| The immunogen recognized by this antibody maps to a region between residue 1175 and 1225 of human Zinc Finger CCCH-type Containing 13 using the numbering given in entry NP_055885.2 (GeneID 23091). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23091 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction