missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562798
This item is not returnable.
View return policy
Description
Immunogen sequence: LILKAISEAQ ESVTKTTNYS TVPQKQTLPV APRTRTSQEE LLAEVVQGQS RTPRISPPIK EEETKGDSVE KNQGTQQRQL LSRLQIDPVM A Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
ZC3H14 belongs to the ZC3H14 family.It contains 2 C3H1-type zinc fingers. ZC3H14 binds the polyadenosine RNA oligonucleotides.
Specifications
| ZC3H14 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H14 | |
| 1010001P15Rik; 1700016A15Rik; 2700069A02Rik; AU014748; FLJ11806; Mammalian suppressor of tau pathology-2; MSUT-2; Npuk68; nuclear protein UKp68; nuclear protein UKp83/UKp68; NY-REN-37; Renal carcinoma antigen NY-REN-37; SUT2; UKp68; Zc3h14; Zinc finger CCCH domain-containing protein 14; zinc finger CCCH type containing 14; zinc finger CCCH-type containing 14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PJT7 | |
| ZC3H14 | |
| Recombinant protein corresponding to Human ZC3H14. Recombinant protein control fragment (Product #RP-103704). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction