missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZC3H15 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16516051
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16516051 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16516051 Supplier Invitrogen™ Supplier No. PA587593

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MPPKKQAQAG GSKKAEQKKK EKIIEDKTFG LKNKKGAKQQ KFIKAVTHQV KFGQQNPRQV AQSEAEKKLK KDDKKKELQE LNELFKPVVA AQKISKGADP KSVVCAFFKQ GQCTKGDKCK FSHDLTLERK CEKRSVYIDA RDEELEKDTM DNWDEKKLEE VVNKKHGEAE KKKPKTQIVC KHFLEAIENN KYGWFWVCPG GGDICMYRHA LPPGFVLKKD KKKEEKEDEI SLEDLIERER SALGPNVTKI TLESFLAWKK RKRQEKIDKL EQDMERRKAD FKAGKALVIS GREVFEFRPE; Positive Samples: HL-60, SH-SY5Y, LO2, Jurkat, MCF7, Mouse liver; Cellular Location: Cytoplasm, Nucleus.

Protects DRG1 from proteolytic degradation (PubMed:19819225). Stimulates DRG1 GTPase activity likely by increasing the affinity for the potassium ions (PubMed:23711155). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3H15
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Zc3h15
Gene Accession No. Q3TIV5, Q8WU90
Gene Alias 1700006A17Rik; 1810012H02Rik; 2610312B22Rik; brain zinc finger protein; DFRP1; DRG family-regulatory protein 1; epo-immediate response gene protein FM22; FM22; HSPC303; HT010; Ierepo4; LEREPO4; likely ortholog of mouse immediate early response erythropoietin 4; MSTP012; p48ZnF; PP730; Zc3h15; Zinc finger CCCH domain-containing protein 15; zinc finger CCCH-type containing 15
Gene Symbols Zc3h15
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZC3H15 (NP_0609412).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55854, 69082
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.