missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H18-AS1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00400555-B01P-50ug
This item is not returnable.
View return policy
Description
ZC3H18-AS1 Polyclonal antibody specifically detects ZC3H18-AS1 in Human samples. It is validated for Western Blot
Specifications
| ZC3H18-AS1 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| Chromosome 16 Open Reading Frame 85, FLJ45530, LINC00859, Long Intergenic Non-Protein Coding RNA 859 | |
| C16orf85 (AAI53154.1, 1 a.a. - 147 a.a.) full-length human protein. MIVSRYGLVGCDAGQVRPKVELSLPAFLALPKKEFKDKPEVEENNLIEEAVWQTSLKIPLPQAQVRERGGGSAGISAAHPSQLCTAGVRPSSALRASRPGWSLQTAALQEPTGADSFLPRSHKTAVDLHSARIPLYIHTKCLLMESV | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAI53154.1 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 400555 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction