missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554862
This item is not returnable.
View return policy
Description
Immunogen sequence: GQLQPSRPTR ARGTCSVEDP LLVCQKEPGK PRMVKSVGSV GDSPREPRRT VSESVIAVKA SFPSSALPPR TGVALGRKLG SHSVASCAPQ LLGDRRVGAG H Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 66%.
Required for the export of polyadenylated mRNAs from the nucleus (PubMed:19364924). Enhances ACVR1B-induced SMAD-dependent transcription. Binds to single-stranded DNA but not to double-stranded DNA in vitro. Involved in RNA cleavage. [UniProt].
Specifications
| ZC3H3 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H3 | |
| BC049953; Kiaa0150; Smad-interacting CPSF-like factor; Smicl; ZC3H3; Zc3hdc3; Zfp623; Zinc finger CCCH domain-containing protein 3; zinc finger CCCH type containing 3; zinc finger CCCH type domain containing 3; zinc finger CCCH-type containing 3; zinc finger CCCH-type domain containing 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23144 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IXZ2 | |
| ZC3H3 | |
| Recombinant protein corresponding to Human ZC3H3. Recombinant protein control fragment (Product #RP-93724). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction