missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570082
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Guinea Pig: 86%; Horse: 91%; Human: 100%; Mouse: 100%; Rat: 100%.
Required for the export of polyadenylated mRNAs from the nucleus (PubMed:19364924). Enhances ACVR1B-induced SMAD-dependent transcription. Binds to single-stranded DNA but not to double-stranded DNA in vitro. Involved in RNA cleavage. [UniProt].
Specifications
| ZC3H3 | |
| Polyclonal | |
| Unconjugated | |
| ZC3H3 | |
| BC049953; Kiaa0150; Smad-interacting CPSF-like factor; Smicl; ZC3H3; Zc3hdc3; Zfp623; Zinc finger CCCH domain-containing protein 3; zinc finger CCCH type containing 3; zinc finger CCCH type domain containing 3; zinc finger CCCH-type containing 3; zinc finger CCCH-type domain containing 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 223642 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8CHP0 | |
| ZC3H3 | |
| synthetic peptide corresponding to a region of mouse ZC3H3 (aa 4-53). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction