missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3H7A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540447
This item is not returnable.
View return policy
Description
Peptide sequence: NSFSMSESKR DLSTSTSREG TPLNNSNSSL LLMNGPGSLF ASENFLGISS Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 93%.
ZC3H7A contains 1 C2H2-type zinc finger, 3 C3H1-type zinc fingers and 3 TPR repeats. The function of ZC3H7A remains unknown.
Specifications
| ZC3H7A | |
| Polyclonal | |
| Unconjugated | |
| ZC3H7A | |
| A430104C18Rik; AI447294; AW556219; FLJ20318; HSPC055; ZC3H7; ZC3H7A; ZC3HDC7; Zinc finger CCCH domain-containing protein 7A; Zinc finger CCCH domain-containing protein 7A-like protein; zinc finger CCCH type containing 7 A; zinc finger CCCH type domain containing 7; zinc finger CCCH-type containing 7A; zinc finger CCCH-type domain containing 7; zinc-finger protein AY163807 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 29066 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IWR0 | |
| ZC3H7A | |
| Synthetic peptide directed towards the middle region of human ZC3H7A. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction