Learn More
ZC3H8 Antibody (OTI4H5), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-74908AF405
Product Code. 18129575
  • € 751.00 / 0.10mL
Description
ZC3H8 Monoclonal specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZC3H8
Monoclonal
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
ZC3H8
Full length recombinant protein of human ZC3H8 (NP_115883) produced in E.coli.
0.1 mL
84524.0
Store at 4°C in the dark.
Immunohistochemistry
OTI4H5
Immunohistochemistry
Fliz1, ZC3HDC8, zinc finger CCCH domain-containing protein 8, zinc finger CCCH type domain containing 8, zinc finger CCCH-type containing 8, zinc finger CCCH-type domain containing 8
Mouse
Affinity Purified