Learn More
ZC3H8 Monoclonal Antibody (OTI5B12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808242S
Encompass_Preferred
 View more versions of this product
Product Code. 17549283
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
ZC3H8 Monoclonal antibody specifically detects ZC3H8 in Human samples. It is validated for Western Blot
Specifications
Specifications
ZC3H8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8N5P1
Mouse
Affinity chromatography
RUO
84524
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5B12
Unconjugated
ZC3H8
AU020882; E130108N08Rik; fetal liver zinc finger 1; fetal liver zinc finger 1; putative zinc finger protein FLIZ1; fetal liver zinc finger protein 1; Fliz1; ZC3H8; Zc3hdc8; zinc finger CCCH domain-containing protein 8; zinc finger CCCH type containing 8; zinc finger CCCH type domain containing 8; zinc finger CCCH-type containing 8; zinc finger CCCH-type domain containing 8
Full length human recombinant protein of human ZC3H8 produced in E.coli.
30 μL