missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H8 Monoclonal Antibody (OTI9F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808290S
This item is not returnable.
View return policy
Description
ZC3H8 Monoclonal antibody specifically detects ZC3H8 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ZC3H8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N5P1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 84524 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI9F11 | |
| Unconjugated | |
| ZC3H8 | |
| AU020882; E130108N08Rik; fetal liver zinc finger 1; fetal liver zinc finger 1; putative zinc finger protein FLIZ1; fetal liver zinc finger protein 1; Fliz1; ZC3H8; Zc3hdc8; zinc finger CCCH domain-containing protein 8; zinc finger CCCH type containing 8; zinc finger CCCH type domain containing 8; zinc finger CCCH-type containing 8; zinc finger CCCH-type domain containing 8 | |
| Full length human recombinant protein of human ZC3H8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction