missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H8 Monoclonal Antibody (OTI9F11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808290
This item is not returnable.
View return policy
Description
ZC3H8 acts as a transcriptional repressor of the GATA3 promoter. Sequence-specific DNA-binding factor that binds to the 5′-AGGTCTC-3′ sequence within the negative cis-acting element intronic regulatory region (IRR) of the GATA3 gene. Component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III. Induces thymocyte apoptosis when overexpressed, which may indicate a role in regulation of thymocyte homeostasis.Specifications
| ZC3H8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8N5P1 | |
| ZC3H8 | |
| Full length human recombit protein of human ZC3H8 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI9F11 | |
| Unconjugated | |
| ZC3H8 | |
| Fliz1, ZC3HDC8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84524 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction