missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZC3HAV1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106389
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZC3HAV1.
The zinc finger antiviral protein (ZC3HAV1) is a CCCH type zinc finger protein that induces an innate immunity to infections by retrovirus by preventing the accumulation of viral RNAs in the cytoplasm and recruits the RNA processing exosome to degrade target RNAs, thereby inhibiting virus replication. ZC3HAV1 is localized in the cytoplasm at steady state, but shuttles between nucleus and cytoplasm in a XPO1-dependent manner. ZAP is a direct target gene of IRF3 action in cellular antiviral responses.
Specifications
| ZC3HAV1 | |
| Polyclonal | |
| Unconjugated | |
| Zc3hav1 | |
| 1200014N16Rik; 2900058M19Rik; 9130009D18Rik; 9830115L13Rik; ADP-ribosyltransferase diphtheria toxin-like 13; ARTD13; CCCH-type zinc finger antiviral protein (Zap); D6Bwg1452e; FLB6421; Inactive Poly [ADP-ribose] polymerase 13; PARP13; PRO1677; rZAP; ZAP; ZC3H2; Zc3hav1; ZC3HDC2; zinc finger antiviral protein; Zinc finger CCCH domain-containing protein 2; zinc finger CCCH type, antiviral 1; zinc finger CCCH-type antiviral protein 1; zinc finger CCCH-type containing, antiviral 1; zinc finger CCCH-type, antiviral 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 252832, 56829, 78781 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3UPF5, Q7Z2W4, Q8K3Y6 | |
| Zc3hav1 | |
| A synthesized peptide derived from human ZC3HAV1(Accession Q7Z2W4), corresponding to amino acid residues A288-D338. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction