missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZCCHC14 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548612
This item is not returnable.
View return policy
Description
ZCCHC14 Polyclonal Antibody for Western Blot
ZCCHC14 gene ontology annotations related to this gene include nucleic acid binding; phosphatidylinositol binding; zinc ion binding.
Specifications
| ZCCHC14 | |
| Polyclonal | |
| Unconjugated | |
| ZCCHC14 | |
| AA792890; BDG29; BDG-29; BDG-29 protein; KIAA0579; Zcchc14; Zinc finger CCHC domain-containing protein 14; zinc finger CCHC-type containing 14; zinc finger, CCHC domain containing 14 | |
| Rabbit | |
| Protein A, Antigen affinity chromatography | |
| RUO | |
| 142682, 23174 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8VIG0, Q8WYQ9 | |
| ZCCHC14 | |
| KLH conjugated synthetic peptide between 420-449 amino acids from the Central region of human ZCCHC14. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction