missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase III/CA3 Antibody (006), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89538APC
This item is not returnable.
View return policy
Description
Carbonic Anhydrase III/CA3 Monoclonal antibody specifically detects Carbonic Anhydrase III/CA3 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Carbonic Anhydrase III/CA3 | |
| Monoclonal | |
| APC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 006 | |
| Western Blot, Immunoprecipitation | |
| CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Carbonic Anhydrase III/CA3 (Accession#: NP_005172.1; Met1-Lys260). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 761 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction