missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHC17 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 196.00 - € 468.00
Specifications
| Antigen | ZCCHC17 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ZCCHC17 Polyclonal antibody specifically detects ZCCHC17 in Mouse samples. It is validated for Western BlotSpecifications
| ZCCHC17 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 51538 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| HSPC251, nucleolar protein of 40 kDa, Pnn-interacting nucleolar protein, pNO40nucleolar protein 40, PS1D protein, PS1DRP11-266K22.1, putative S1 RNA binding domain protein, Putative S1 RNA-binding domain protein, Zinc finger CCHC domain-containing protein 17, zinc finger, CCHC domain containing 17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-241 of human ZCCHC17 (NP_057589.2). MNSGRPETMENLPALYTIFQGEVAMVTDYGAFIKIPGCRKQGLVHRTHMSSCRVDKPSEIVDVGDKVWVKLIGREMKNDRIKVSLSMKVVNQGTGKDLDPNNVIIEQEERRRRSFQDYTGQKITLEAVLNTTCKKCGCKGHFAKDCFMQPGGTKYSLIPDEEEEKEEAKSAEFEKPDPTRNPSRKRKKEKKKKKHRDRKSSDSDSSDSESDTGKRARHTSKDSKAAKKKKKKKKHKKKHKE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title