missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZCCHC18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5139962
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VVSLYPSLTP TGAPPFRGRA RPLDQVL Highest antigen sequence indentity to the following orthologs: Mouse - 41%.
ZCCHC18 is a protein coding gene.
Specifications
| ZCCHC18 | |
| Polyclonal | |
| Unconjugated | |
| ZCCHC18 | |
| 1500031H04Rik; LOW QUALITY PROTEIN: zinc finger CCHC domain-containing protein 18; paraneoplastic Ma antigen family member 7B; PNMA7B; putative zinc finger CCHC domain-containing protein 18; Sizn2; SMAD-interacting zinc finger protein 2; Zcchc18; Zinc finger CCHC domain-containing protein 18; zinc finger CCHC-type containing 18; zinc finger, CCHC domain containing 12 pseudogene 1; zinc finger, CCHC domain containing 18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 644353 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0CG32 | |
| ZCCHC18 | |
| Recombinant protein corresponding to Human ZCCHC18. Recombinant protein control fragment (Product #RP-109676). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction