missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZCCHC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA545115
This item is not returnable.
View return policy
Description
Peptide sequence: KNRNWEKHRK ADRHREVDED FPRGPKTYSS PGSFKTQKPS KPFHRSSHYH Sequence homology: Horse: 86%; Human: 100%; Pig: 79%.
The function of this protein remains unknown.
Specifications
| ZCCHC7 | |
| Polyclonal | |
| Unconjugated | |
| ZCCHC7 | |
| 4930572I07Rik; AIR1; D4Wsu132e; HSPC086; TRAMP-like complex RNA-binding factor ZCCHC7; ZCCHC7; Zinc finger CCHC domain-containing protein 7; zinc finger CCHC-type containing 7; zinc finger, CCHC domain containing 7 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 84186 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N3Z6 | |
| ZCCHC7 | |
| Synthetic peptide directed towards the C-terminal of human ZCCHC7 (aa 453-502). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction