missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHV Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77184AF700
This item is not returnable.
View return policy
Description
ZC3HAV1L Polyclonal antibody specifically detects ZC3HAV1L in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ZC3HAV1L | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 700 | |
| C7orf39, chromosome 7 open reading frame 39, MGC14289, zinc finger CCCH-type antiviral protein 1-like, zinc finger CCCH-type, antiviral 1-like | |
| Antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human ZC3HAV1. The immunogen is located within amino acids 680 - 730 of ZC3HAV1. Amino Acid Squence: QMKRGPDHQPAKTS | |
| 0.1 mL | |
| Innate Immunity | |
| 92092 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction