Learn More
Invitrogen™ ZCWPW1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596731
Description
Immunogen sequence: VMKKRRNDCS QKLGVALMMA QEAEQISIQE RVNLFGFWSR FNGSNSNGER KDLQLSGLNS PGSCLEKKEK EEELEKEEGE KTDPILPIRK RVKIQTQKTK PRGLGGDAGT ADGRGRTLQR KIMKRSLGRK STAPPAPRMG RKEGQGNSDS DQPGPKKKFK APQSKALAAS FSEGKEVRTV PKNLGLSACK GACPSSAKEE PRHREPLTQE AGSVPLEDEA SSDLDLEQLM EDVGRELGQS GELQHSNSDG EDFPVALFGK; Positive Samples: SW480, BT-474, HepG2, Mouse brain.
Specifications
| ZCWPW1 | |
| Polyclonal | |
| Unconjugated | |
| ZCWPW1 | |
| Gm1053; ZCW1; Zcwpw1; zinc finger CW type with PWWP domain 1; zinc finger CW-type and PWWP domain 1; zinc finger CW-type and PWWP domain containing 1; zinc finger CW-type PWWP domain protein 1; zinc finger, CW type with PWWP domain 1; zinc finger, CW-type with PWWP domain 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 381678, 55063 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.07 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6IR42, Q9H0M4 | |
| ZCWPW1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 389-648 of human ZCWPW1 (NP_0604543). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.