missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZDHHC1 Polyclonal Antibody for ELISA, ICC/IF, IHC (P), Western Blot
ZDHHC1 (Zinc Finger DHHC-Type Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include protein-cysteine S-palmitoyltransferase activity and palmitoyltransferase activity.
Specifications
Specifications
| Antigen | ZDHHC1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.4 |
| Gene | ZDHHC1 |
| Gene Accession No. | Q8R0N9, Q8WTX9 |
| Gene Alias | 4432412D04Rik; C16orf1; DHHC domain-containing cysteine-rich protein 1; DHHC-1; DHHC-domain-containing cysteine-rich protein; HSU90653; probable palmitoyltransferase ZDHHC1; ZDHHC1; Zinc finger DHHC domain-containing protein 1; zinc finger DHHC-type containing 1; zinc finger protein 377; zinc finger, DHHC domain containing 1; zinc finger, DHHC-type containing 1; ZNF377 |
| Gene Symbols | ZDHHC1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
