Learn More
ZDHHC11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-15166
Product Code. 18705893
  • € 621.00 / 0.10mL
Description
ZDHHC11 Polyclonal specifically detects ZDHHC11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.