missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP3-38335-20ul
Les retours ne sont pas autorisés pour ce produit.
Consulta la politica di reso
Descrizione
ZDHHC17 Polyclonal antibody specifically detects ZDHHC17 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifica
| ZDHHC17 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA | |
| DHHC-17, EC 2.3.1, EC 2.3.1.-, HIP14HIP-14, huntingtin interacting protein 14, huntingtin interacting protein 3, Huntingtin interacting protein H, Huntingtin yeast partner H, Huntingtin-interacting protein 14, Huntingtin-interacting protein 3, Huntingtin-interacting protein H, HYPHHIP-3, KIAA0946HIP3, palmitoyltransferase ZDHHC17, Putative MAPK-activating protein PM11, Putative NF-kappa-B-activating protein 205, Zinc finger DHHC domain-containing protein 17, zinc finger, DHHC domain containing 17, zinc finger, DHHC-type containing 17 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 170-310 of human ZDHHC17 (NP_056151.2).,, Sequence:, TSIVAYLIAKGQDVDMMDQNGMTPLMWAAYRTHSVDPTRLLLTFNVSVNLGDKYHKNTALHWAVLAGNTTVISLLLEAGANVDAQNIKGESALDLAKQRKNVWMINHLQEARQAKGYDNPSFLRKLKADKEFRQKVMLGTP | |
| 20 μL | |
| Signal Transduction | |
| 23390 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto