missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC17 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68744
This item is not returnable.
View return policy
Description
ZDHHC17 Polyclonal antibody specifically detects ZDHHC17 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| ZDHHC17 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| DHHC-17, EC 2.3.1, EC 2.3.1.-, HIP14HIP-14, huntingtin interacting protein 14, huntingtin interacting protein 3, Huntingtin interacting protein H, Huntingtin yeast partner H, Huntingtin-interacting protein 14, Huntingtin-interacting protein 3, Huntingtin-interacting protein H, HYPHHIP-3, KIAA0946HIP3, palmitoyltransferase ZDHHC17, Putative MAPK-activating protein PM11, Putative NF-kappa-B-activating protein 205, Zinc finger DHHC domain-containing protein 17, zinc finger, DHHC domain containing 17, zinc finger, DHHC-type containing 17 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LGITTNERMNARRYKHFKVTTTSIESPFNHGCVRNIIDFFEFRCCGLFRPVIVDWTRQYTIEYDQISGSGYQL | |
| 100 μg | |
| Signal Transduction | |
| 23390 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction