missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93085-0.1ml
This item is not returnable.
View return policy
Description
ZDHHC2 Polyclonal antibody specifically detects ZDHHC2 in Human, Rat samples. It is validated for Western Blot
Specifications
| ZDHHC2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| DHHC2, DHHC-2, EC 2.3.1, EC 2.3.1.-, palmitoyltransferase ZDHHC2, Ream, REC, Reduced expression associated with metastasis protein, Reduced expression in cancer protein, Zinc finger DHHC domain-containing protein 2, Zinc finger protein 372, zinc finger, DHHC domain containing 2, zinc finger, DHHC-type containing 2, ZNF372rec | |
| A synthetic peptide corresponding to a sequence within amino acids 270-367 of human ZDHHC2 (NP_057437.1). QVFGDEKKYWLLPIFSSLGDGCSFPTCLVNQDPEQASTPAGLNSTAKNLENHQFPAKPLRESQSHLLTDSQSWTESSINPGKCKAGMSNPALTMENET | |
| 0.1 mL | |
| Cancer, GPCR, Signal Transduction | |
| 51201 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction