missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZDHHC20 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17279293 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17279293 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17279293 Supplier Invitrogen™ Supplier No. PA5116796

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Samples: HeLa, MCF7, SKOV3, A-549, Mouse brain, Mouse lung, Mouse testis, Rat brain, Rat lung Immunogen sequence: VFGDEKKYWL LPIFSSLGDG CSFPTRLVGM DPEQASVTNQ NEYARSSGSN QPFPIKPLSE SKNRLLDSES QWLENGAEEG IVKSGTNNHV TVAIEN.

Catalyzes palmitoylation of Cys residues on target proteins (PubMed:27153536, PubMed:29326245). Catalyzes palmitoylation of Cys residues in the cytoplasmic C-terminus of EGFR, and modulates the duration of EGFR signaling by modulating palmitoylation-dependent EGFR internalization and degradation (PubMed:27153536). Has a preference for acyl-CoA with C16 fatty acid chains (PubMed:29326245). Can also utilize acyl-CoA with C14 and C18 fatty acid chains (PubMed:29326245). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZDHHC20
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZDHHC20
Gene Accession No. Q5W0Z9, Q5Y5T1
Gene Alias 4933421L13Rik; 5033406L14Rik; AI448102; B230110O18Rik; DHHC-20; DHHC-containing protein 20; HK11; membrane-associated DHHC24 zinc finger protein; Palmitoyltransferase ZDHHC20; probable palmitoyltransferase ZDHHC20; RGD1305755; ZDHHC20; Zinc finger DHHC domain-containing protein 20; zinc finger DHHC-type containing 20; zinc finger, DHHC domain containing 20; zinc finger, DHHC-type containing 20
Gene Symbols ZDHHC20
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 270-365 of human ZDHHC20 (NP_0013169881).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253832, 305923, 75965
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.