missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 238.00 - € 481.00
Specifications
| Antigen | ZDHHC4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680711
|
Novus Biologicals
NBP2-94486-0.02ml |
0.02 mL |
€ 238.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603161
|
Novus Biologicals
NBP2-94486-0.1ml |
0.1 mL |
€ 481.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZDHHC4 Polyclonal antibody specifically detects ZDHHC4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ZDHHC4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| DHHC domain containing 4, DHHC-4, EC 2.3.1, EC 2.3.1.-, Zinc finger protein 374, zinc finger, DHHC-type containing 4 | |
| A synthetic peptide corresponding to a sequence within amino acids 110-210 of human ZDHHC4 (NP_060576.1). VNLFFFTLTCGTNPGIITKANELLFLHVYEFDEVMFPKNVRCSTCDLRKPARSKHCSVCNWCVHRFDHHCVWVNNCIGAWNIRYFLIYVLTLTASAATVAI | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 55146 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title