missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116794
This item is not returnable.
View return policy
Description
Positive Samples: HeLa, Rat lung Immunogen sequence: VNLFFFTLTC GTNPGIITKA NELLFLHVYE FDEVMFPKNV RCSTCDLRKP ARSKHCSVCN WCVHRFDHHC VWVNNCIGAW NIRYFLIYVL TLTASAATVA I.
ZDHHC4 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include integral component of membrane; protein-cysteine S-palmitoyltransferase activity.
Specifications
| ZDHHC4 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC4 | |
| 1810021D01Rik; 2900029I10Rik; DC1; DHHC-4; membrane-associated DHHC4 zinc finger protein; probable palmitoyltransferase ZDHHC4; UNQ5787/PRO19576; Zdhhc4; Zinc finger DHHC domain-containing protein 4; zinc finger DHHC-type containing 4; zinc finger protein 374; zinc finger, DHHC domain containing 4; zinc finger, DHHC-type containing 4; ZNF374 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 304291, 55146, 72881 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.25 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5FVR1, Q9D6H5, Q9NPG8 | |
| ZDHHC4 | |
| A synthetic peptide corresponding to a sequence within amino acids 110-210 of human ZDHHC4 (NP_0605761). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction