Learn More
Novus Biologicals™ ZDHHC5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30004
This item is not returnable.
View return policy
Description
ZDHHC5 Polyclonal antibody specifically detects ZDHHC5 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ZDHHC5 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DHHC domain containing 5, DKFZp586K0524, zinc finger, DHHC-type containing 5, ZNF375 | |
| Between 500 and 550 | |
| 100 μg | |
| Cell Biology | |
| 25921 | |
| Store at 4°C. Do not freeze. | |
| IgG |