missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZDHHC8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 292.00 - € 572.00
Specifications
| Antigen | ZDHHC8 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18410361
|
Novus Biologicals
NBP1-81418-25ul |
25ul |
€ 292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18257875
|
Novus Biologicals
NBP1-81418 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZDHHC8 Polyclonal specifically detects ZDHHC8 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZDHHC8 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DHHC8, EC 2.3.1, EC 2.3.1.-, KIAA1292DHHC-8, membrane-associated DHHC8 zinc finger protein, probable palmitoyltransferase ZDHHC8, ZDHHCL1, Zinc finger DHHC domain-containing protein 8, Zinc finger protein 378, zinc finger, DHHC-type containing 8, ZNF378zinc finger, DHHC domain like containing 1 | |
| ZDHHC8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 29801 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PKIEAGTFSSDLQTPRPGSAESALSVQRTSPPTPAMYKFRPAFPTGPKVPFCGPGEQVPGPDSLTLGDDSIRSLDFVSEPSLDLPDYGPGGLHAAYPPSPPLSASDAFSGALRSLSLKASSRRGGDHVAL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title