missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-13159AF594
This item is not returnable.
View return policy
Description
ZEB1 Polyclonal specifically detects ZEB1 in Human, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ZEB1 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| AREB6MGC133261, delta-crystallin enhancer binding factor 1, DELTAEF1, FECD6, Negative regulator of IL2, NIL2A, NIL-2-A, NIL-2-A zinc finger protein, posterior polymorphous corneal dystrophy 3, PPCD3, TCF-8, TCF8BZP, Transcription factor 8, transcription factor 8 (represses interleukin 2 expression), ZEB, Zfhep, ZFHX1A, zinc finger E-box binding homeobox 1, zinc finger E-box-binding homeobox 1, zinc finger homeodomain enhancer-binding protein | |
| Rabbit | |
| 0.1 mL | |
| 6935 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| ZEB1 | |
| A synthetic peptide made to an C-terminal portion of the human ZEB1 protein (between residues 1087-1124). [Uniprot: P37275] | |
| Primary | |
| Human, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction