missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB2 Antibody (923328) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB73782JF669
This item is not returnable.
View return policy
Description
ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| ZEB2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b | |
| E. coli-derived recombinant human ZEB2/SIP1, Asn363-Lys537, Accession # O60315 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), CyTOF | |
| 923328 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 9839 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction