missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 17 publications
Brand: Novus Biologicals NBP1-82991-25ul
This item is not returnable.
View return policy
Description
ZEB2 Polyclonal specifically detects ZEB2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| ZEB2 | |
| Polyclonal | |
| Western Blot 0.04 - 0.4 ug/ml, Flow Cytometry, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated | |
| KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human ZEB2 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZEB2 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SPVKPMDSITSPSIAELHNSVTNCDPPLRLTKPSHFTNIKPVEKLDHSRSNTPSPLNLSSTSSKNSHSSSYTPNSFSSEELQAEPLDLSLPKQMKEPKSIIATKNKTKASSISLDHNSVSSSSE | |
| 25 μL | |
| Core ESC Like Genes, Neuroscience, Stem Cell Markers | |
| 9839 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction