Learn More
ZEB2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 17 publications
Brand:  Novus Biologicals NBP1-82991
 View more versions of this product
Product Code. 18729663
  • € 572.00 / 0.10mL
Description
ZEB2 Polyclonal specifically detects ZEB2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, Flow Cytometry, Immunohistochemistry
Western Blot 0.04 - 0.4 ug/ml, Flow Cytometry, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000, Knockdown Validated
IgG
Polyclonal
KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b