missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZEB2 Monoclonal Antibody (OTI4G3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802114
This item is not returnable.
View return policy
Description
ZEB2, initially identified as Smad interacting-protein 1, is normally located in the nucleus and functions as a DNA-binding transcriptional repressor that interacts with activated SMADs. Like the homologous ZEB1, ZEB2 inhibits the transcription of the E-cadherin gene and induces epithelial-mesenchymal transition, a genetic program controlling cell migration during embryonic development and wound healing, in vitro. ZEB2 can also protect cells from DNA damage-induced apoptosis, suggesting that its expression may contribute to tumor progression. Recent evidence has shown that ZEB2 is often observed in the cytoplasm in numerous cancer tissues, indicating that its localization may be regulated in normal and tumor tissues. Mutations in this gene are also associated with Hirschsprung disease/Mowat-Wilson syndrome.Specifications
| ZEB2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60315 | |
| ZEB2 | |
| Human recombit protein fragment corresponding to amino acids 1-350 of human ZEB2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI4G3 | |
| Unconjugated | |
| ZEB2 | |
| HSPC082, SIP-1, SIP1, SMADIP1, ZFHX1B | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 9839 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction