missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Basement Membrane Marker Antibody (FIS 5F11/2), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50369DL594
This item is not returnable.
View return policy
Description
Zebrafish Basement Membrane Marker Monoclonal antibody specifically detects Zebrafish Basement Membrane Marker in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| Zebrafish Basement Membrane Marker | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| FIS 5F11/2 | |
| 50 mM Sodium Borate | |
| Zebrafish Intestine | |
| 0.1 mL | |
| Developmental Biology, Epitope Tags | |
| Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction