Learn More
Zebrafish Basolateral Pole of Cells Antibody (FIS 2H9/1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP2-50370CL1
Product Code. 30157239
  • € 898.00 / 0.10mL
Description
Zebrafish Basolateral Pole of Cells Monoclonal antibody specifically detects Zebrafish Basolateral Pole of Cells in Zebrafish samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zebrafish Basolateral Pole of Cells
Monoclonal
CoraFluor 1
Mouse
Protein A purified
RUO
Zebrafish
Purified
Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
FIS 2H9/1
PBS
Lysate of zebrafish intestine
0.1 mL
Primary
Store at 4°C in the dark. Do not freeze.