missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Basolateral Pole of Cells Antibody (FIS 2H9/1), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50370H
This item is not returnable.
View return policy
Description
Zebrafish Basolateral Pole of Cells Monoclonal specifically detects Zebrafish Basolateral Pole of Cells in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Zebrafish Basolateral Pole of Cells | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Lysate of zebrafish intestine | |
| 0.1 mL | |
| Developmental Biology, Epitope Tags | |
| Zebrafish | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FIS 2H9/1 | |
| Western Blot, Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction