missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Delta C Antibody (zdc2), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50371APCCY7
This item is not returnable.
View return policy
Description
Zebrafish Delta C Monoclonal specifically detects Zebrafish Delta C in Zebrafish samples. It is validated for Flow Cytometry.
Specifications
| Zebrafish Delta C | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| Extracellular region of Zebrafish Delta C protein | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| zdc2 | |
| Flow Cytometry | |
| Mouse | |
| Protein A purified | |
| Cell Cycle and Replication, Developmental Biology, Neuroscience | |
| Zebrafish | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction