missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Delta C Antibody (zdc2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 201.00 - € 351.00
Specifications
| Antigen | Zebrafish Delta C |
|---|---|
| Clone | zdc2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18655007
|
Novus Biologicals
NBP2-50371-0.025ml |
0.025 mL |
€ 201.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674878
|
Novus Biologicals
NBP2-50371 |
0.1 mL |
€ 351.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zebrafish Delta C Monoclonal antibody specifically detects Zebrafish Delta C in Zebrafish samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, CyTOFSpecifications
| Zebrafish Delta C | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Developmental Biology, Neuroscience | |
| PBS | |
| IgG2a | |
| Protein A purified |
| zdc2 | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Zebrafish | |
| Extracellular region of Zebrafish Delta C protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title