missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Delta D Antibody (zdd2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50372H
This item is not returnable.
View return policy
Description
Zebrafish Delta D Monoclonal specifically detects Zebrafish Delta D in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| Zebrafish Delta D | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| Extracellular domain; Zebrafish Delta D peptide | |
| 0.1 mL | |
| Cell Cycle and Replication, Developmental Biology, Neuroscience | |
| Specific for Zebrafish Delta D extracellular domain. This antibody does not detect Delta C, but it cross-reacts weakly with Delta A. | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry | |
| zdd2 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction