missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 2F11/2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50375F
This item is not returnable.
View return policy
Description
Zebrafish Gut Secretory Cell Epitopes Monoclonal specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Zebrafish Gut Secretory Cell Epitopes | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Lysate of zebrafish intestine | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
| Zebrafish | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| FIS 2F11/2 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction