missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Gut Secretory Cell Epitopes Antibody (FIS 6G5/1), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50377DL594
This item is not returnable.
View return policy
Description
Zebrafish Gut Secretory Cell Epitopes Monoclonal antibody specifically detects Zebrafish Gut Secretory Cell Epitopes in Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Zebrafish Gut Secretory Cell Epitopes | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| FIS 6G5/1 | |
| 50mM Sodium Borate | |
| Lysate of zebrafish intestine | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Signaling, Developmental Biology, Epigenetics, Epitope Tags | |
| Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction