Learn More
ZER1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H9G7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-68258-20UL
 View more versions of this product
Product Code. 18049885
  • € 145.00 / 20µL
Description
The ZER1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZER1. This antibody recognizes human antigen. The ZER1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
ZER1
1H9G7
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG2a
Immunofluorescence
1000 μg/mL
C9orf60, Hzyg, Protein zer 1 homolog, RP11 545E17.4, zer 1 homolog (C. elegans), ZER1, ZYG, Zyg 11 homolog B like protein, ZYG11BL
20 μL
10444
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.