Learn More
ZER1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H9G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68258-20UL
Description
The ZER1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZER1. This antibody recognizes human antigen. The ZER1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| ZER1 | |
| 1H9G7 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence | |
| 1000 μg/mL | |
| C9orf60, Hzyg, Protein zer 1 homolog, RP11 545E17.4, zer 1 homolog (C. elegans), ZER1, ZYG, Zyg 11 homolog B like protein, ZYG11BL | |
| 20 μL | |
| 10444 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |