Learn More
ZVG Zellstoff Vertrieb™ Zetmask FFP2 NR D Filter Mask with Exhalation Valve
Brand: ZVG Zellstoff Vertrieb™ 3097201
Additional Details : Weight : 0.25000kg
Specifications
EN 149: 2002 | |
Filter Mask | |
Protection Against Slightly Toxic and Harmful Substances, | |
Yes |
Single-use | |
Yes | |
Exhalation valve | |
10/Pk. |