missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFAND2B Antibody (OTI1G10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74917APC
This item is not returnable.
View return policy
Description
ZFAND2B Monoclonal specifically detects ZFAND2B in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| ZFAND2B | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| ZFAND2B | |
| Full length recombinant protein of human ZFAND2B (NP_620157) produced in HEK293T cells. | |
| 0.1 mL | |
| 130617.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI1G10 | |
| Flow Cytometry | |
| AIRAP-like protein, AN1-type 2B, AN1-type zinc finger protein 2B, arsenite inducible RNA associated protein-like, Arsenite-inducible RNA-associated protein-like protein, zinc finger, AN1-type domain 2B | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction