Learn More
ZFAND2B Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA502398S
Encompass_Preferred
 View more versions of this product
Product Code. 17526801
  • € 242.00 / 30µL
Description
ZFAND2B Monoclonal antibody specifically detects ZFAND2B in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
ZFAND2B
Monoclonal
0.43 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8WV99
Mouse
Affinity chromatography
RUO
130617
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Western Blot
OTI2E4
Unconjugated
ZFAND2B
1110060O18Rik; AIRAPL; AIRAP-like protein; AN1-type zinc finger protein 2B; arsenite-inducible RNA-associated protein-like protein; C81256; ZFAND2B; zinc finger AN1-type containing 2B; zinc finger, AN1 type domain 2B; zinc finger, AN1-type domain 2B
Full length human recombinant protein of human ZFAND2B produced in HEK293T cell.
30 μL